Would you like to log in again?
Automated Zebrafish 5 Choice Chamber
Supplier: Maze Engineers
4871
76670-128EA 7986.67 USD InStock
76670-128
Automated Zebrafish 5 Choice Chamber
Animal Supplies Animal Environmental Enrichment
The Zebrafish choice chamber allows for experiments similar to the commonly used five-choice serial reaction time task (5-CSRTT).
  • Acrylic five-choice maze with five apertures
  • Adjustable feeding slide to control the feeding amount
  • Easy to clean
During testing, lights are illuminated and the gate is raised. A food delivery apparatus delivers rewards at a fixed time schedule. The proper light is illuminated in the interval or until the correct entry is chosen. The number of trials, accuracy, omissions, and latency are usually recorded. This choice chamber includes an automated gate, lights on the proximal and distal ends, and control of lights and gate using conductor software.
Ordering information: The automated feeder is designed for use with the automated Zebrafish choice chamber. The accessory software package is designed for use with the automated Zebrafish five choice chamber as a stand alone software package.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty