Would you like to log in again?
Automated Zebrafish T-Maze
Supplier: Maze Engineers
4876
76670-138EA 9320 USD InStock
76670-138
Automated Zebrafish T-Maze
Animal Supplies Animal Environmental Enrichment
The Zebrafish Automated T-Maze is used to study spatial learning and memory, along with conditioned behaviors in Zebrafish.
  • Acrylic construction
  • Easy to clean
The Zebrafish Automated T-Maze consists of a chamber with test areas and returning pathways. It features seven auto doors and sensor arrays, two reward dispensers, two auto water stirrers, control box and other accessories.
Ordering information: The Automated Zebrafish T-Maze requires software to function (see accessory options). Software package is designed for use with the Automated Zebrafish T-Maze as a standalone software package.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty