Would you like to log in again?
Resident Intruder Tests
Supplier: Maze Engineers
4904 4903
76670-146EA 1986.67 USD InStock
76670-146 76670-148
Resident Intruder Tests
Animal Supplies Animal Environmental Enrichment
The Resident Intruder test is a chamber used for social defeat paradigms.
  • Clear and white acrylic chamber
  • Metal mesh top
  • Easy to clean
Composed of a clear cage front for video recording, opaque sides and back, wire top, and a bottom lip to keep in bedding. Easy to clean by spraying and wiping with 70% ethanol, without losing clarity of front wall.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty