Would you like to log in again?
Social Defeat Apparatus
Supplier: Maze Engineers
4905 4906
76670-152EA 2386.67 USD InStock
76670-152 76670-150
Social Defeat Apparatus
Animal Supplies Animal Environmental Enrichment
The social defeat test allows for study of recurrent defeat and is often used in the assessment of psychiatric compounds.
  • Clear acrylic construction
  • Apparatus includes one arena and two mesh enclosures
Repetitive exposure encourages anhedonia, social avoidance behaviors, and anxiety, and although relatively new (developed in 2006), is seen as a simple yet powerful model of social interaction. The social defeat apparatus comes with two social defeat cages, an arena, and a clear divider for use in the home cage. Additional cages or modifications can be made upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty