Would you like to log in again?
Social Reward Chambers
Supplier: Maze Engineers
4907 4908
76670-156EA 1853.34 USD InStock
76670-156 76670-154
Social Reward Chambers
Animal Supplies Animal Environmental Enrichment
The social reward chamber is designed to understand the effect of social rewards on the social preferences of the subject.
  • Clear acrylic construction
Unlike the existing models that are exclusively based on the Conditioned Place Preference (CPP) paradigm and require memory association (Trezza et al., 2011), the Social Reward Chamber is based on Operant Social Preference (OSP) tasks that take advantage of visual stimulus (Borland et al., 2017).
The social reward chamber for mouse features a main chamber measuring 33×22×20 cm, and two additional chambers measuring 17×12×20 cm. The social reward chamber for rat features a main chamber measuring 50×33×30 cm, and two additional chambers measuring 25×18×30 cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty