Would you like to log in again?
Burrowing Tubes
Supplier: Maze Engineers
4916 4917 4920 4915 4919 4918
76670-168EA 386.67 USD InStock
76670-168 76670-166 76670-176 76670-174 76670-172 76670-170
Burrowing Tubes
Animal Supplies Animal Environmental Enrichment
The Burrowing tube studies burrowing behaviour in which rodents spontaneously empty a tube filled with food pellets, gravel, or other substances. This is a sensitive test to various pathologies including prion diseases and inflammatory conditions.
  • Acrylic construction
  • Non reflective surfaces
  • Easy to clean
The Burrowing tube is manufactured from acrylic, with acrylic tubing and a 1 cm fiberboard backing that is sealed with a waterproof adhesive. Each tube includes legging on the proximal side for elevation off of the floor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty