Would you like to log in again?
Horizontal and Vertical Grid Test
Supplier: Maze Engineers
5003 5001 5002 5004
76670-188EA 920 USD InStock
76670-188 76670-186 76670-184 76670-182
Horizontal and Vertical Grid Test
Animal Supplies Animal Environmental Enrichment
The horizontal grid apparatus allows for the study of neuromuscular strength and coordination in rodents. The vertical grid test is similar to the horizontal grid test to assess motor function, neuromuscular strength, and coordination.
  • Acrylic frame construction with metal mesh
  • Simple to use and easy to clean, assemble, and disassemble
The horizontal grid apparatus is a horizontally positioned grid box mounted with plexiglass, which allows the subect to grab onto the grids as it climbs across while upside down. The vertical grid apparatus is a vertically positioned grid box mounted with acrylic, and this allows the subject to grab on the grids as it climbs down.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty