Would you like to log in again?
Gait Tests
Supplier: Maze Engineers
5008 5009
76670-198EA 2653.34 USD InStock
76670-198 76670-196
Gait Tests
Animal Supplies Animal Environmental Enrichment
The Gait test is a useful apparatus to measure motor lesions and other dysfunctions related to gait.
  • Simple and reliable
  • Easy to clean
In the Gait test, the forepaw of the rodent is wetted with blue black ink (included) and placed on one end of the runway, which is covered with a strip of paper. The stride length of forelimbs are measured manually as the distance between two forepaw prints. This apparatus comes with an optional lighting device to encourage motivation to the dark housing area. Sizing for mice and rats available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty