Would you like to log in again?
Body Turning Tests
Supplier: Maze Engineers
5012 5011
76670-202EA 653.34 USD InStock
76670-202 76670-204
Body Turning Tests
Animal Supplies Animal Environmental Enrichment
The body turning test is a narrow alleyway that allows mice and rats to navigate turning or reversing.
  • Acrylic construction
  • Non reflective surfaces
  • Removable walls for easy cleaning and storage
A narrow corridor open at one end and closed at the other (three walls), all resting on a removable base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty