Would you like to log in again?
Gap Cross Tests
Supplier: Maze Engineers
5027 5026
76670-230EA 2520 USD InStock
76670-230 76670-228
Gap Cross Tests
Animal Supplies Animal Environmental Enrichment
The Gap Cross Test is a behavioral test used to assess a subject’s ability to navigate a gap or void in a platform.
  • Acrylic construction
  • Non-reflective surfaces
  • Easy cleaning and maintenance
The Gap Cross Test is typically used to study anxiety and fear-related behaviors, as well as motor coordination and balance. It is a commonly used tool in behavioral neuroscience research to study anxiety and related behaviors, and can be used to study the effects of various treatments, such as drugs or manipulations of the brain, on these behaviors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty