Would you like to log in again?
Horizontal Ladder Tests
Supplier: Maze Engineers
5102 5101 5103 5104
76670-242EA 566.67 USD InStock
76670-242 76670-240 76670-238 76670-236
Horizontal Ladder Tests
Animal Supplies Animal Environmental Enrichment
The horizontal ladder test is a motor and coordination test for evaluating skilled walking for rodents.
  • Stainless steel rungs, acrylic frame and tub
  • Two acrylic tubs included with full package formats
  • E, easy to assemble, disassemble, and clean
In the horizontal ladder test, steps are counted and classified as either correct/functional paw placement, a slip, or a miss. Fore and hind-limb coordination can be assessed further by removing individual ladder rungs to evaluate how the rodent targets individual rungs. This model either includes the ladder and two tubs or the ladder alone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty