Would you like to log in again?
Geotaxis Tests, Acrylic or Stainless Steel
Supplier: Maze Engineers
5105 5107 5106 5108
76670-246EA 2653.34 USD InStock
76670-246 76670-244 76670-250 76670-248
Geotaxis Tests, Acrylic or Stainless Steel
Animal Supplies Animal Environmental Enrichment
The geotaxis test is used to investigate motor coordination and vestibular sensitivity in rodents.
  • Acrylic construction, for placement at incremental angles
  • Strainless steel construction, for placement at continuous angles
  • Easy to clean
The rodent is usually placed in an inclined grid in the head downwards position. Available in acrylic or stainless steel models.
Acrylic models offer placement at angles of 45, 40, 35, 30, 25, 20, 15, 10, or 5°. Stainless steel models provide constant angulation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty