Would you like to log in again?
Sleep Deprivation Apparatus
Supplier: Maze Engineers
5603
76670-292EA 6653.34 USD InStock
76670-292
Sleep Deprivation Apparatus
Animal Supplies Animal Environmental Enrichment
Sleep deprivation (SD) impairs spatial, emotional, and working memories, and augments anxiety-like behaviors.
  • Chamber dimensions: 28D×25H cm
  • Time and rotation apparatus dimensions: 25L×10W×25H cm
  • Lid slots allow for cables and cannulas, for applications in optogenetics, EEG, and more
The rodent sleep deprivation chamber apparatus is an automated device that provides movement at researcher programmed intervals to prevent REM sleep. The apparatus consists of a clear acrylic chamber atop a motor and screen to set time and rotations of a bar inside the chamber to prevent the subject from sleeping in a safe way. Food and water apparatuses are available to allow long-term experimentation. Lid slots allow for cable and cannula use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty