Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rotarod Apparatus

Supplier: Maze Engineers
5703 5704
76670-298EA 7060 USD InStock
76670-298 76670-296
Rotarod Apparatus
Animal Supplies Animal Environmental Enrichment
The rotarod apparatus is widely used as a performance and behavioral task to assess motor performance in rodents using the natural fear of falling as motivation.

  • Angular acceleration: 0.1 to 50 rpm
  • Acceleration time range: 1 to 4999 seconds
  • Angular speed: 0.1 to 99.9 rpm
  • Change in increments of 0.1 rpm

The Rotarod device can be customized for speed and acceleration, and is connected and controlled using the easy-to-integrate Conductor software (included). Dividers prevent mice from entering other lanes. Minimal height and a soft landing prevents harm after a fall from the bar. The rod itself has horizontal ridges to help the rodent hold onto the rod.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty