Would you like to log in again?
Self-Administration Chambers
Supplier: Maze Engineers
5842 5841
76670-320EA 10920 USD InStock
76670-320 76670-318
Self-Administration Chambers
Animal Supplies Animal Environmental Enrichment
The self-administration chamber is used to assess reward-seeking and addiction behaviors. This chamber offers everything needed for operant-led self-administration assessments associated with food restriction and intravenous drug administration.
  • Rodent chamber with two nose poke apertures or levers
  • Two cue lights in the apertures
  • One pellet dispenser, infusion pump, frequency tone, shock grid, and shocker (optional)
Data analysis which can be performed with the self-administration chamber include: number of infusions; number of active lever presses or nose-pokes; number of inactive lever presses or nose-pokes.
The chamber comes with a sound-attenuating cubicle with light or fan, a syringe pump system, a cage with two portlights and retractable levers, a shock floor grid, and two pellet receptacles. The option for an optic sensor to detect nose pokes can be added.
Ordering information: The Self-administration chamber requires software to function (see accessory option). Software package is designed for use with the Self administration apparatus as standalone software.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty