Would you like to log in again?
Active/Passive Avoidance Shuttle Boxes
Supplier: Maze Engineers
6002 6001
76670-326EA 10533.34 USD InStock
76670-326 76670-324
Active/Passive Avoidance Shuttle Boxes
Animal Supplies Animal Environmental Enrichment
The shuttle box is a flexible system for both active and passive avoidance experiments.
  • Acrylic plating allows for adjustable contexts
  • Black/black plating for active avoidance
  • White/black plating for passive avoidance
  • Sound Attenuation Chamber sold separately
The Shuttle box inclues two independent grid floors that allow for flexible adverse stimuli. A top-loading door allows easy access inside the box. The cage contains a sound generator and a visual stimulus (light) that functions separately for each compartment.
Rodents in the cage are detected by two separate sensor arrays that have high sensitivity and specificity. Data can be combined with the Noldus Ethovision software and integrated with Neuralynx using the Conductor software.
Ordering information: The Active/Passive Avoidance Shuttle Box requires software to function (see accessory option). Software package is designed for use with the Active/Passive apparatus.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty