Would you like to log in again?
5-Choice Serial Reaction Time Tasks
Supplier: Maze Engineers
6101 6102
76670-354EA 9320 USD InStock
76670-354 76670-352
5-Choice Serial Reaction Time Tasks
Animal Supplies Animal Environmental Enrichment
The 5-Choice Serial Reaction Time Task (5CSRTT) was developed to study behavioral traits associated with attention deficit hyperactivity disorder (ADHD), such as attention and impulse control.
  • Includes five nose poke apertures
  • Feces and urine tray is removable
  • Aluminum back wall Isolation cubicle not included
The 5CSRTT apparatus consists of an operant conditioning chamber with two Plexiglas sidewalls. The aluminum front wall is rounded and contains five nose poke apertures (2.5×2.2×2.2 cm each, 2 cm above the floor). Each apparatus comes with a stimulus LED light as well as an infrared nose poke sensor. A stainless steel back wall has a food cachet installed which provides food pellets from the pellet dispenser. The shocker floor grid allows for punishment in the experiment. The removable feces and urine tray below this grid allow for easy cleaning.
Ordering information: Software package (see accessory option) is designed for use with the automated 5CSRTT as stand alone software.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty