Would you like to log in again?
Parallel Rod Floor Tests
Supplier: Maze Engineers
6502 6504 6503 6501
76670-378EA 1186.67 USD InStock
76670-378 76670-376 76670-382 76670-380
Parallel Rod Floor Tests
Animal Supplies Animal Environmental Enrichment
The parallel rod floor test was originally used as a simple model of ataxia in rodents.
  • Clear acrylic and metal grid
  • Easy to clean, assemble, and disassemble
The parallel rod floor test was originally described in the literature by Kamans and Crabbe (2007) as a simple model of ataxia in mice. The test allows researchers to quantify differences in motor coordination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty