Would you like to log in again?
Multivariate Concentric Square Field Tests
Supplier: Maze Engineers
6602 6601
76670-386EA 3453.34 USD InStock
76670-386 76670-384
Multivariate Concentric Square Field Tests
Animal Supplies Animal Environmental Enrichment
The Multivariate Concentric Square Field test (MCSF) is a novel behavior model for investigation of risk assessment, anxiety, and shelter seeking behavior in rodents.
  • Non-reflective acrylic surfaces
  • Easy to clean
The MCSF was originally described in the literature by Meyerson et al., 2006, for rats and later adopted by Ekmark-Lewen for mice. The behavioral model provides several areas for the animal to explore by free choice, including sheltered, open, and elevated areas.
The MCSF mouse model features a 72×72 cm open field area, with a 42×42 cm central chamber. The MCSF rat model features a 95×95 cm open field area, with a 55×55 cm central chamber.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty