Would you like to log in again?
Chronic Social Defeat Stress Cages
Supplier: Maze Engineers
6702 6701
76670-388EA 1720 USD InStock
76670-388 76670-390
Chronic Social Defeat Stress Cages
Animal Supplies Animal Environmental Enrichment
The chronic social defeat stress cage is used for testing depression in rodents, using a chronic social defeat stress model.
  • Clear acrylic construction
  • Easy to clean
The chronic social defeat stress task is performed to impose both physical and sensory stress to the subject through social defeat sessions with an aggressor, followed by a period of sensory stress.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty