Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hot/Cold Plate Test

Supplier: Maze Engineers
8012
76670-406EA 6660 USD InStock
76670-406
Hot/Cold Plate Test
Animal Supplies Animal Environmental Enrichment
The hot/cold Plate test serves as a useful screening tool for interventions of analgesia in rodents.

  • Digital temperature display mode
  • Temperature range: 20 to 65 °C
  • Temperature precision: 0.01 °C
  • Temperature control stdev: <±0.2 °C
  • Time display range: >10 hours
  • Low, medium, and high warm-up time options

The hot/cold plate test is a gold standard thermal pain test in rodents and serves as a useful screening tool for interventions of analgesia. The rodent is placed into a chamber with a heated floor with surrounding clear acrylic walls. Two key behaviors are measured; paw licking and jumping. The hot/cold plate apparatus is composed of a transparent glass cylinder and a hot/cold plate. The plantar surface is a metal floor equipped with adjustable temperature control. The glass cylinder is used to limit the subject to the plate and prevent it from jumping off the apparatus while allowing an unobstructed view. The hot/cold plate test combines both cold and hot stimulus apparatus into a single unit. The cold plate test allows an understanding of animal sensitivity to cold temperatures that enables the assessment of cold-induced hyperalgesia and allodynia. The apparatus allows testing of unilateral pain quantification using the cold plate which isn’t possible with the hot plate test alone. The increase or decrease in the rate of cooling and heating allows observation of thermal stimulus temperatures that incite nocifensive responses. The apparatus is effective in the assessment of the effectiveness of analgesics.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty