Would you like to log in again?
OroFacial Pain Assessment Device (OPAD)
Supplier: Maze Engineers
8013
76670-408EA 7866.67 USD InStock
76670-408
OroFacial Pain Assessment Device (OPAD)
Animal Supplies Animal Environmental Enrichment
The Orofacial pain assessment device (OPAD) is an operant system of pain assessment that relies on voluntary behavior. Assays using the OPAD offer a unidimensional assessment of pain.
  • Clear acrylic cage with a metal base
  • Custom dimensions available
  • Temperature range from lab room temperature: 25 to 75° C
The Orofacial Pain Assessment Device (OPAD) was developed by Neubert et al., (2005) as an operant system of pain assessment that relies on voluntary behavior. Responses involve executive functioning and other experiences, thus relying simply on reflex and innate responses does not provide the complete picture. Therefore, a conflict-based paradigm proves to be a more sensitive method that enables in-depth analysis of pain. The OPAD system is composed of a thermode and metal wires that allow assessment of thermal and mechanical sensitivity. The starved subject is placed in the operant chamber where it can access the food reward only when it contacts the thermal and mechanical stimuli. Essentially, the subject is tasked with choosing to tolerate the stimuli to gain the reward. Unlike traditional assays that are typically based on reflex behaviors, the OPAD allows the subject to choose its own stimuli threshold in order to attain the reward. This reward-conflict paradigm offers better face, content, and predictive validity.
Ordering information: Software package (see accessory option) is designed for use with the Orofacial assay as standalone software.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty