Would you like to log in again?
Drosophila Y-Maze
Supplier: Maze Engineers
8502
76670-414EA 1320 USD InStock
76670-414
Drosophila Y-Maze
Animal Supplies Animal Environmental Enrichment
The Drosophila Y-Maze is a laboratory task used to study spatial learning and memory in Drosophila melanogaster.
  • Acrylic construction
  • Ten sets of 10 Y-Mazes
In this task, the fruit fly is placed at the end of one arm of a Y-shaped maze and is allowed to freely explore the maze. It is a simple and widely used experimental paradigm in which a fly is placed in a Y-shaped maze and given the opportunity to choose between two arms of the maze. The fly’s choice is recorded and the fly is then given a reward, such as food, in one of the arms. The fly is then placed back in the maze and given the opportunity to choose again.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty