Would you like to log in again?
Drosophila Shallow Chambers
Supplier: Maze Engineers
8506 8507
76670-418EA 2120 USD InStock
76670-418 76670-420
Drosophila Shallow Chambers
Animal Supplies Animal Environmental Enrichment
The shallow drosophila chamber was first described by Simon et al (2010) as an innovative design to restrict flies to a shallow volume of space, forcing all behavioral interactions to take place within a monolayer of individuals.
  • 7 cm model is available for courtship and aggression studies
  • 13 cm model is available for large groups of drosophila
  • 11 degree sloped flooring, glass ceiling, LED multi-light display
  • Visual stimulus of a 12×12" array of 850 nm IR LED mounted underneath the chambers for backlighting
The chamber design minimizes the frequency that flies occlude or obscure each other, and promotes a greater number of flies to move throughout the center of the chamber. This increases the frequency of interactions and improves the quality of data collected by digital video.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty