Would you like to log in again?
Drosophila Gravitaxis Maze
Supplier: Maze Engineers
8508
76670-422EA 2653.34 USD InStock
76670-422
Drosophila Gravitaxis Maze
Animal Supplies Animal Environmental Enrichment
The Drosophila gravitaxis maze is used to observe positive or negative gravitaxis movement in Drosophila species.
  • Acrylic construction
  • Easy to clean
Drosophila navigate the maze by choosing either to walk upward or downward to choose their preferred exit. The number of up or down choices made by the flies in the maze is used to deduce the gravitaxis behavior of Drosophila.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty