Would you like to log in again?
Caterpillar Y-Maze
Supplier: Maze Engineers
9101
76670-424EA 2920 USD InStock
76670-424
Caterpillar Y-Maze
Animal Supplies Animal Environmental Enrichment
The Caterpillar Y-Maze is shaped to assess memory retention during metamorphosis.
  • Acrylic construction
  • Easy to clean
The Caterpillar Y-Maze features two symmetrical arms and one longitudinal section. It was created in an experiment by Douglas J. Blackiston et al. (2008), to asess memory retention during metamorphosis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty