Would you like to log in again?
VWR® Stackable Mini Rollers
Supplier: VWR
VWR®
MR-01UA-VWR MR-03UA-VWR
76670-460EA 1415.93 USD InStock
76670-460 76670-462
VWR® Stackable Mini Rollers
Roller Culture Apparatus
VWR® Stackable mini rollers, designed for gentle agitation of multiple sized bottles/tubes in a space saving footprint.
  • Adjustable digitial speed control, timer
  • Sealed electronics, constructed of corrosion resistant materials, suitable for use in CO₂/95% Humidity environments
  • Remote control (wired) included, to control unit without disrupting CO₂ Incubator chamber environment
  • 3D swing model (76670-460) offers 18 mm (0.71") swing motion
  • Easy to add and remove rollers to accommodate a wide variety of tube/bottle sizes
  • Optional stacking kit to stack 2 units, maximzing bench space
All models include O-rings for 50 ml tubes and 15 ml tubes for smooth rolling and wired remote control module.
Certifications: CE, CSA.
Delivery information: Supplied with 76670-454, 76670-452, 76670-450, 2 year warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty