Would you like to log in again?
AI Bacteria Analysis Slides
Supplier: Apacor
255001 255000
76670-670PK 23.4 USD InStock
76670-670 76670-672
AI Bacteria Analysis Slides
Slides Microscope Slides
Fiducial printed glass microscope slides for AI image analysis.
  • Printed fiducial design
  • White glass
  • Cut edges
Slides are available in two formats, with either a circle center or square center.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty