Would you like to log in again?
Tempo iMono™ Human iPSC derived CD14+ Monocytes
Supplier: TEMPO BIOSCIENCE, INC
Tempo-iMono™
211
76683-004EA 950.25 USD InStock
76683-004
Tempo iMono™ Human iPSC derived CD14+ Monocytes
Assays Cellular Assays
Human iPSC-derived CD14+ Monocytes.
  • Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty