Would you like to log in again?
ActivArmr® 96-002 Goatskin Leather Gloves
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ActivArmr®
RIGLVCVR080 RIGLVCVR070 RIGLVCVR110 RIGLVCVR100 RIGLVCVR120 RIGLVCVR090
76683-112CS 202.07 USD InStock
76683-112 76683-132 76683-120 76683-128 76683-138 76683-114
ActivArmr® 96-002 Goatskin Leather Gloves
Gloves
Goatskin leather protectors, for enhanced low-voltage glove life and improved hand safety. Designed to be worn over Class 0 and 00 electrical safety gloves, these leather protectors can significantly extend wear life.
  • Extend the life of rubber insulated gloves while providing excellent hand protection
  • Provides cut, puncture, and abrasion protection
  • Manufactured to ASTM D120 and EN 60903 standards
  • Suitable to wear with Class 0 and 00 class gloves
ActivArmr® 96-002 protectors are made from premium goatskin leather, enhancing hand safety when conducting electrical work with (or close to) energized components, while offering moderate abrasion, puncture, and cut resistance. The leather formulation also offers excellent dexterity, for genuinely easy-to-use electrical PPE.
Certifications: ASTM F496, ASTM F969, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, ISO 9001.
Packaging: Gloves are individually packaged by pair.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty