Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, GemPure™ Select USP for irrigation (WIR)

Supplier: GeminiBio
Synonyms: Dihydrogen oxide

901-003-021 901-003-5X6 901-003-203 901-003-5X6 901-003-021 901-003-1X6 901-003-1X6 901-003-203
76683-178CS 95.86 USD InStock
76683-178 CA76683-184 76683-184 CA76683-182 76683-182 CA76683-180 76683-180 CA76683-178
Water, GemPure™ Select USP for irrigation (WIR)
Water
GeminiBio’s Water for Irrigation (WFIr) Quality Water (USP Grade) is a hypotonic, sterile non-pyrogenic irrigating water produced by our proprietary water purification system. Manufactured and tested in compliance with the USP monograph for bulk packaged water for irrigation, GemPure™ Select Water for Irrigation (WFIr) Quality Water (USP Grade) is ideal for use in the manufacture and testing of medical devices, and applications where Water For Injection is not required.

  • Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
  • Available in 500 ml, 1, 20 and 200 L, as well as custom sizes to suit your needs
  • For further manufacturing or research use only
  • Not for direct therapeutic use

Meets the USP monograph for Water for Irrigation packaged in bulk for commercial use.


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
UN: 2201
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty