Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, GemPure™, Cell and Molecular Grade

Supplier: GeminiBio
Synonyms: Dihydrogen oxide

901-004-1X6 901-004-1X6
76683-186CS 102.02 USD InStock
CA76683-186 76683-186
Water, GemPure™, Cell and Molecular Grade
Water
GeminiBio’s Cell and Molecular Grade Water is manufactured specifically for your most stringent research applications. GemPure™ Cell and Molecular Grade Water is sterile and low endotoxin for your cell culture processes. Protease and nuclease free, it is perfect for any of your molecular biology needs as well saving valuable lab space and simplifying your purchasing process.

  • Tested for RNase, DNase, Protease, sterility, TOC, endotoxin, and more
  • Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
  • Available in 1 L bottle and custom sizes to suit your needs
  • For further manufacturing or research use only
  • Not for direct therapeutic use


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
UN: 2201
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty