Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Mini Stirrer, 230 V (Export Only)

Supplier: VWR
VWR®
30881774 80863874
76683-462EA 292.51 USD InStock
76683-462 10153-316
VWR® Mini Stirrer, 230 V (Export Only)
Stirrers Magnetic Stirrers
Rugged, compact stirrers with an ergonomic design are ideal for educational labs and can stir up to 1000 ml of water.

  • Rugged 5×5" aluminum top plate
  • Speed range of 100 to 1200 rpm
  • Built-in support rod holder
  • Five-Year warranty on parts and labor

Durable 5×5" cast aluminum top plate will not chip or crack.

The Auto Stirrers are ideal for applications that require repeatable results. These compact, powerful stirrers automatically start when a vessel is placed on the aluminum top plate (minimum weight of approximately 50 grams [1.75 oz.]). When the vessel is removed the stirring stops, but the speed setting remains unchanged. Units save power and provide an extra measure of safety by running only when needed. Optional support plate allows stirrer to be mounted to lab frames or ring stands.

The built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Kit includes 45.7 cm (18") support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector. Optional support plate allows hot plate to be mounted to lab frames or ring stands.

Powerful motor and magnet deliver reliable and consistent stirring.

Certifications: TUV listed. CE marked. RoHS certified.

Ordering information: The 230 V units include a 183 cm (72") cord with a Euro style plug. All units include a five-year limited warranty on parts and labor.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty