Would you like to log in again?
VWR® Advanced 3750 Reciprocating Shakers, 230 V
Supplier: VWR
VWR®
80863522
76683-468EA 3488.03 USD InStock
76683-468
VWR® Advanced 3750 Reciprocating Shakers, 230 V
Shakers and Mixers Reciprocating Shakers
Digital reciprocating shakers are designed for a wide range of applications including solubility studies, extraction procedures, and cell cultures that require accurate and reproducible results.
  • 230 V model
  • Exceptional speed control, accuracy, and durability
  • Displays for speed and time
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
The shaking system delivers exceptional speed control, accuracy, and durability while continuously monitoring shaking speed and setpoint, even under changing loads.
Safety features include a built-in load sensor that detects unbalanced conditions and automatically reduces rpm or stops the operation to protect samples. A ramping feature slowly increases speed to the setpoint to avoid splashing. In timed mode, an audible alarm will sound when the timer reaches zero. Shakers can be run in cold rooms, incubators, and CO₂ environments from −10 to +60°C (−14 to +140°F). An RS-232 serial port is standard on these shakers.
Shakers can accommodate a variety of stainless steel platform options. Adjustable platforms are equipped with four cushioned rollers to hold most glassware securely. Universal platforms are suitable for mounting flask clamps and test tube racks. The culture platforms with mat are designed for slow speed applications that utilize low profile vessels. The separatory funnel platform can hold up to three 500 ml to 2 L samples at once. Standard test tube racks are made from PVC coated steel. Pivoting test tube racks are are made from polypropylene in a stainless steel holder.
The variable speed microprocessor control provides consistent, uniform mixing action. The microprocessor will display the last setpoint and will restart the unit if power is interrupted. Permanently lubricated ball bearings and a maintenance-free, brushless DC motor ensure reliable service in continuous operations. Touchpad controls feature easy-to-read, independent LEDs for speed and time, allowing operators to view all settings at once. The timer will display elapsed time or, when programmed to user defined limits, will shut off the unit. Last settings will be displayed even after the power has been shut off.
Certifications: RoHS certified. TUV listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Shaker is supplied with a nonskid rubber mat to fit the 28×33 cm (11×13") tray, designed to secure Petri dishes and cell culture flasks. Unit also includes a 2.3 m (92") detachable 3-wire cord and plug. 230 V, 50/60 Hz models are also available; contact your Avantor sales representative for more information.
Caution: 230 V model. Corresponding models with voltage suitable for use in US (97007-742 or 10027-166) are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty