Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 95%

Supplier: Ambeed
Warning
Synonyms: N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochlorideN1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochlorideTMPD 2 HClN,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

A414780-25G A414780-5G A414780-1G A414780-100G A414780-1G A414780-5G
103827-282EA 7 USD InStock
103827-282 76841-898 76841-900 76694-908 76694-906 103838-326
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 95%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Caution: for Research Use Only


Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Boiling Pt: 260.6 °C (760 mm Hg)
Melting Pt: 220…224 °C at N/A mmHg
Flash Pt: 104.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty