Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Secretoglobin Family 3A Member 2(SCGB3A2) ELISA Kit

Supplier: AFG Bioscience
EK731590
76706-670EA 397.1 USD InStock
76706-670
Mouse Secretoglobin Family 3A Member 2(SCGB3A2) ELISA Kit
Assays ELISAs
  • Detection wavelength: 450 nm

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty