Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human 5-Hydroxytryptamine Receptor 2A(HTR2A) ELISA Kit

Supplier: AFG Bioscience
EK712695
76711-606EA 397.1 USD InStock
76711-606
Human 5-Hydroxytryptamine Receptor 2A(HTR2A) ELISA Kit
Assays ELISAs
  • Detection wavelength: 450 nm

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty