Would you like to log in again?
Lighthouse Launderable Microfiber Mops
Supplier: Unimed
Lighthouse
THCT521122 TMMP446L18 BHCT48977E
76716-784CS 328.68 USD InStock
76716-784 77007-790 77007-792
Lighthouse Launderable Microfiber Mops
Mops
The launderable microfiber mop can be used both as a dry and wet mop to clean floors in any facility or environment.
  • Effective in removing dirt and bacteria
  • Saves money and resources
  • Eco-friendly
  • Can be washed and reused
Microfiber mops can remove dirt and grime from floors more effectively than traditional mops because the fibers attract and hold onto dirt particles. Washable microfiber mops can be used for hundreds of cleanings before needing to be replaced, which saves money on purchasing new mops and reduces waste.
Ordering information: Mop frame and adjustable handle sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty