Would you like to log in again?
ThinkLite Purilux Vent 2×2' LED Light Panel
Supplier: Unimed
ThinkLite
TPRV1039PRO
76716-790EA 2631.38 USD InStock
76716-790
ThinkLite Purilux Vent 2×2' LED Light Panel
Lights LED Lights, Lamps and Bulbs
Purilux Vent 2×2' LED Light Panel
  • Easy to install; fits like a 2×2' troffer/panel
  • Vibration- and impact-resistant
  • Quiet performance
  • Emergency battery backup option available
  • Instant on/off
Originally developed in partnership with epidemiologists at Boston Children’s Hospital, the Purilux silently produces the purest, safest air possible, while also providing a modern, energy efficient lighting solution. The air in buildings equipped with the Purilux is free from airborne pathogens, viruses, mold, environmental pollution, allergens, and other irritants for maximum safety and wellness. Built-in ULPA and carbon filters team up with a medical grade, centrifugal fan and a UVC/UVA sanitizing chamber to purify the air, eliminating viruses and other airborne risks, including TVOCs, without the use of any chemicals, ions, or ozone.
Studies show that clean, healthy indoor air significantly lessens the transmission of disease, boosts focus and productivity for workers, improves cognitive function, leads to less absenteeism due to illness, reduces the impact of asthma and allergies, supports deep, restful sleep, and lowers the risk of respiratory illness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty