Would you like to log in again?
Lighthouse Canister Wall Bracket
Supplier: Unimed
Lighthouse
TMWB446000
76716-816EA 15.88 USD InStock
76716-816
Lighthouse Canister Wall Bracket
Wipe Dispensers
Holds surface wipe canisters securely and conveniently on mobile work stations.
  • Heavy-duty ³/₁₆" thick steel with white powdercoat finish for maximum durability
  • Integral antimicrobial coating
  • Mounts on the lower, wider pole of cart
  • Can be mounted to the upper pole with the undermounting kit
Bracket is designed to accommodate a surface wipe canister, for use on mobile work stations. The heavy-duty ³/₁₆" thick steel with white powdercoat finish provides maximum durability, and features an integral antimicrobial coating.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty