Would you like to log in again?
Lighthouse Wire Glove Box Holders
Supplier: Unimed
Lighthouse
BHQH004060 BHDH004040 BVTH004055 BVDH004045 BHTH004050 BXWW004029 BVQH004065
76716-852EA 19.73 USD InStock
76716-852 76716-774 76716-810 76716-818 76716-828 76716-856 76716-858
Lighthouse Wire Glove Box Holders
Glove Dispensers and Glove Box Holders
Economical organizer keeps gloves clean and ready to use. Accommodates most brands and sizes of glove boxes.
  • Open design allows view of sizes and glove types
  • Steel construction with baked-on white finish
  • Includes wall mounting hardware
Multiple glove box holders allow for OSHA compliance by dispensing sizes and glove types (latex or nitrile) for all staff needs. Economical for use at industrial sites, nurses offices, patient rooms, doctor’s offices, laboratories, EMS areas, etc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty