Would you like to log in again?
Sapphire FL Biomolecular Imager
Supplier: Azure Biosystems
IS4007 IS4004 IS4001 IS4005 IS4059 IS4027 IS4058 IS4006 IS4003 IS4055 IS4000 IS4002 IS4056 IS4057
76717-006EA 14746.05 USD InStock
76717-006 77449-126 77449-138 76716-950 77449-144 76716-952 77449-146 76716-996 77449-140 76716-992 77449-142 76716-988 76716-954 76716-956
Sapphire FL Biomolecular Imager
Photodocumentation Systems
Flexibility with uncompromising performance – from in vitro molecular assays to in vivo imaging.
  • Sapphire FL is the ultimate biomolecular imager for flexibility
  • Laser scanning system designed for western blots, 96 well plates, animal imaging and more
With customizable and user-changeable laser and filter modules, the Sapphire FL easily adapts to a lab’s changing needs and advancing research. The Sapphire FL offers customizable and user-changeable optical modules, 5 to 1000 µm resolution scans, a Z-plane range from −1.0 to +6 mm, 5 anesthesia ports for imaging living animals, chemiluminescence detection through the chemiluminescence module and much more.
Certifications: CE, cTUVca.
Order Now
ORDER
ACCESSORIES
SERVICES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty