Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NEBExpress® T4 Lysozyme
Supplier: New England Biolabs
NEBExpress®
P8115S P8115L
76717-038EA 419 USD InStock
76717-038 76717-042
NEBExpress® T4 Lysozyme
Competent Cells
NEBExpress T4 Lysozyme is 200-fold more active than chicken egg white lysozyme.
  • Lysis reactions are scalable and compatible with high throughput workflows
  • Fast non-mechanical bacterial lysis; the lysate is ready to use and compatible with affinity resins
  • Small (200 µg) and large (5×200 µg) formats suitable for lysing 7 or 35 g of cells, respectively
  • Recombinant, animal free and REACH compliant
NEBExpress T4 Lysozyme is a recombinant murein hydrolase that breaks down the bacterial cell wall by hydrolyzing the beta 1 to 4 linkage between N-acetylmuramic acid and N-acetylglucosamine in the peptidoglycan of prokaryotic cells (gram-negative and some gram-positive bacteria). It can be used to extract soluble proteins, membrane proteins, DNA, RNA, or metabolites.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty