Would you like to log in again?
AccuForge® Single Edge Blades, Coated, Unwrapped
Supplier: AccuTec Blades
AccuForge®
AGBL-7007-0000
76717-298CS 43896 USD InStock
76717-298
AccuForge® Single Edge Blades, Coated, Unwrapped
Razors and Blades
AccuForge single edge blades feature 3-facet edges for sharpness. The MicroCoat® Enrichment technology reduces cut force and increases durability.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • Unwrapped blades are ideal for high volume uses
  • 3-facet edge for increased sharpness
  • Blades are packed in a wooden crate
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty