Would you like to log in again?
AccuForge® Single Edge Blades, 10 Blade Safety Dispenser
Supplier: AccuTec Blades
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AccuForge®
AGBL-7024-0000 AGBL-7008-0000 AGBL-7017-0000 AGBL-7039-0000
76717-312CS 1080 USD InStock
76717-312 76717-334 76717-322 76717-300
AccuForge® Single Edge Blades, 10 Blade Safety Dispenser
Razors and Blades
AccuForge single edge blades feature 3-facet edges for sharpness. Each dispenser holds 10 blades and includes used blade storage.
  • 3-facet edge for increased sharpness
  • Transparent safety dispenser with used blade vault
  • Available degreased to remove residual oil
  • Available with MicroCoat® enrichment technology
  • Each dispenser contains 10 blades
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty