Would you like to log in again?
AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Supplier: AccuTec Blades
AccuForge®
AGBL-7010-0000 AGBL-7009-0000 AGBL-7014-BULK
76717-302CS 857.49 USD InStock
76717-302 76717-306 76717-304
AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Razors and Blades
AccuForge single edge blades feature 3-facet edges for sharpness. Blades are degreased to remove residual oil.
  • 3-facet edge for increased sharpness
  • Degreased to remove oil residue
  • Paper-free packaging is clean room ready
  • Individually packed in clamshell with product key and instructions for use
  • Available in bulk pack without clamshells
  • Each cartridge contains 100 blades
Ordering information: For use with GEM RUNNER™ Dispenser Cat no. 76717-342.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty