Would you like to log in again?
AccuForge® Single Edge Blades, 3 Facet
Supplier: AccuTec Blades
AccuForge®
AGBL-7030-0000 AGBL-7018-0000 AGBL-7019-0000 AGBL-7021-0000
76717-314CS 1056.25 USD InStock
76717-314 76717-318 76717-316 76717-326
AccuForge® Single Edge Blades, 3 Facet
Razors and Blades
AccuForge single edge blades feature 3-facet edges for sharpness. Blades are wrapped in protective paper.
  • 3-facet edge for increased sharpness
  • Blades are individually wrapped for safety
  • Re-closable clamshell for safe and convenient blade storage
  • Each clamshell contains 100 blades
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty