Would you like to log in again?
AimStrip® Xylazine Urine/Powder Dipstick, Germaine Laboratories
Supplier: Germaine Laboratories
AimStrip™
43347
76717-408PK 78.38 USD InStock
76717-408
AimStrip® Xylazine Urine/Powder Dipstick, Germaine Laboratories
Drug Tests
AimStrip® Xylazine Urine/Powder Dipstick, 25/Kit. Cut-off concentration of 1000 ng/ml.
  • XYL has been reported to be misused as a horse doping agent, a drug of abuse and as source of accidental or intended poisonings.
  • Human intoxication with XYL has been reported sporadically over the past several decades
  • Noted in literature describing drug overdose deaths in Philadelphia as early as 2006
  • Since mid-2010's, XYL has been noticed by people who inject drugs (PWID) and public health practitioners as an increasingly commonplace additive in the street opioid supply of Philadelphia.
  • Recent reports from Connecticut implicated XYL in a rising fraction of overdose deaths in 2019–2020
  • In Puerto Rico, heroin is commonly adulterated with XYL
  • Frequently found in speedball (a cocaine and heroin mixture)
A rapid test for the qualitative detection of xylazine in human urine and powder. For forensic use only, not for in vitro diagnostic use. A rapid chromatographic immunoassay for the detection of xylazine in human urine and powders at a cut-off concentration of 1000 ng/ml. The Xylazine (XYL) Rapid Test Dipstick is a rapid screening test that can be performed without the use of an instrument. The test utilizes the antibody to selectively detect elevated levels of xylazine in urine and powders.
XYL has been reported to be misused as a horse doping agent, a drug of abuse and as source of accidental or intended poisonings.
Delivery information: Ships ground, can ship expedited if specified on PO or requested timely with sales@germainelabs.com
Caution: For forensic use only, not for in vitro diagnostic use
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty