Would you like to log in again?
SlideMate™ Slides and SlideMate™ Plus Adhesion Slides
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SlideMate™
TT-8041I-PS-8218-1 TT-5041I-8217-001 TT-6041I-PS-8218-1 TT-7041I-PS-8218-1 TT-4041I-8217-001 TT-7041I-8217-001 TT-4041I-PS-8218-1 TT-8041I-8217-001 TT-6041I-8217-001 TT-5041I-PS-8218-1
76717-420CS 591.7 USD InStock
76717-420 76717-424 76717-412 76717-422 76717-410 76717-416 76406-502 76717-414 76406-500 76717-418
SlideMate™ Slides and SlideMate™ Plus Adhesion Slides
Slides Adhesive Slides
SlideMate™ Microscope Slides designed for use on thermal printers.
  • Manufactured in the USA
  • SlideMate™ slides are smooth, three-quarter inch white tab slides validated to work with the SlideMate™ AS slide printer
  • The tab is impervious to virtually all common laboratory chemicals and reagents
  • Available in adhesion and non-adhesion formulas and in multiple colored tabs
  • SlideMate™ Plus Adhesion Slides feature adhesion coating for routine clinical applications
Experience crisp, clean printing with SlideMate™ microscope slides from Epredia™. Designed for use on SlideMate™ thermal printers. SlideMate™ slides can help reduce identification issues common with handwritten labels. Available in adhesion and non-adhesion varieties.
Epredia™ SlideMate™ slides are made from extra-white glass, manufactured to high quality standards. The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
SlideMate™ slides come pre-cleaned, flat with finely ground edges 90°. Each package comes wrapped to help protect against humidity and moisture. The packaging is also designed to assist with loading the slides into SlideMate™ printer hoppers to prevent contamination due to handling.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty