Would you like to log in again?
Human Anti-Dengue Antibody IgG Titer Serologic Assay Kit (Envelope, E)
Supplier: ReVacc Scientific
P76A64-S1 P76A64-S2 P76A64-COM
76729-566EA 459 USD InStock
76729-566 76729-606 76729-526
Human Anti-Dengue Antibody IgG Titer Serologic Assay Kit (Envelope, E)
Assays Immunological Assays
This kit is developed for serologic test for human IgG titer of Anti-Dengue E antibody in serum/plasma or purified human antibody (monoclonal & polyclonal) in vitro. It is for research use only.
This kit was tested to bind to a published monoclonal antibody (1A1D2) starting from concentration of 10 μg/ml.
Assay Principles: This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-Dengue E human IgG in serum. The kit consists of high-bind detachable 96-well plate pre-coated with protein, Positive control, Negative Control, an HRP-Anti-Human IgG secondary antibody, TMB and dilution/wash/stop buffer. Use protocol (4 simple steps): 1.      Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37°C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml). 2.      Dilute the Secondary antibody HRP-Anti-Human IgG by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5-1 hour. Avoid light by foil. Wash wells as mentioned. 3.      Add TMB to each well (0.1 ml) and incubate at room temperature for 5-30 minutes. 4.      Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD Value reflects the amount of antibody bound. Storage and shipping condition: Shipment with room temperature. The unopened kit is stable for 6 months from the date of manufacture when stored at 2°C to 8°C. Avoid light. The shelf life is 60 days from the date of opening.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty